missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes

Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.

Brand:  Agilent BioTek 7170013

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10676615

  • £1510.00 / Each
Estimated Shipment: 29-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Volume range 1 μL to 10 mL
  • Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
  • Dispense 1536 wells in 18 s.
  • Cassettes can be autoclaved without recalibration
  • Dispense up to eight reagents at one time
  • Customizable dispense pattern allows skipped columns or rows
  • Built-in priming trough
  • Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
  • Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput

Specifications

Specifications

0.014 in.
For liquid handling
0.035 cm
Stainless Steel
10 to 3000 μL
Large Volume Cassette
10 μL
EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers