missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff
GREENER_CHOICE

Product Code. 16429415
Change view
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16429415 X-Large Pack of 100
16499405 Small Pack of 100
16409415 Medium Pack of 100
16419415 Large Pack of 100
16439415 2X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16429415 Supplier Ansell Supplier No. 93862100
Only null left
Add to basket
Edge
Add to basket

Disposable nitrile gloves with extended cuff provide durable protection in the event of chemical splashes. NFPA and medical exam grade certified for use by healthcare professionals.

  • Tested against both fentanyl and gastric acid (vomit) to simulate authentic overdose situations
  • Dark, black color makes it easy to see when hands have come into contact with potentially hazardous powders
  • Made with a soft, flexible nitrile formulation
  • Fully textured for a reliable, consistent grip
  • For use with:
    • Administering drugs
    • Emergency services
    • Equipment repair and maintenance
    • Extra protection over wrist and forearm
    • Inspection, selecting, checking parts
    • Laboratory analysis and testing
    • Paint and spray shops
    • Patient examinations
    • Raw material sample collection
    • Tire rotation and change
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 455 1-4
Accelerator Free No
Applications Healthcare, Laboratory
AQL 0.65
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 6.3 mil
Finger Thickness (Metric) 0.16 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Sterility Non-sterile
Thickness (English) 6.3 mil
Thickness (Metric) 0.16 mm
Model 93862100
Series MICROFLEX™ MidKnight™ XTRA 93-862
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.