missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ LifeStar™ EC 93-868 High Risk Nitrile Exam Gloves

Product Code. 16855440 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
12
Unit Size:
Pack of 100
Product Code. Size unitSize
16855440 8 Pack of 100
16845440 7 Pack of 100
16885440 11 Pack of 100
16895440 12 Pack of 100
16865440 9 Pack of 100
16875440 10 Pack of 100
6 options
This item is not returnable. View return policy

Product Code. 16855440

Brand: Ansell 93868080

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Dual-layered gloves provide two levels of reliable barrier protection against hazardous substances and high-risk patient handling.

  • Two layers of reliable barrier protection
  • For protection against hazardous substances and high-risk patient handling
  • Designed with non-stick and non-foaming properties
  • Reduced interference and increased productivity
  • Advanced barrier protection for demanding tasks
  • AQL 0.65
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 455 1-4, EN 420:2003 + A1:2009
Accelerator Free Yes
Applications Industrial
AQL 0.65
Color White
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 8
Sterility Non-sterile
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKPT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
For Use With (Application) Handling hazardous substance and high-risk patients
Glove Cuff Style Beaded
Series 93-868
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.