missing translation for 'onlineSavingsMsg'
Learn More

Ansell Microflex™ 93-852 Distinctive Black Nitrile Exam Glove

Product Code. 17909241 Shop All Ansell Products
Change view
Click to view available options
Size:
2X-Large
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17909241 2X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17909241 Supplier Ansell Supplier No. 93852110
Stock Up Your Lab
Use promo code 28007
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Non-foaming for easy handling of objects in wet environments. Distinctive black color hides stains and provides contrast for improved visibility. Fully textured design for confident grip.

  • Thanks to its non-foaming special formulation, Microflex 93-852 offers a secure grip in wet applications/environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) Category III|EC 2023/2006|EC 1935/2004|EN 455 1-4|EN ISO 21420:2020|EN ISO 374-1:2016 Type B|EN ISO 374-5:2016|ISO 11193|ISO 18889:2019
Chlorinated No
Accelerator Free No
Applications Industrial, Laboratory
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 5.9 mil
Finger Thickness (Metric) 0.15 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 2X-Large
Sterility Non-sterile
Thickness (English) 4.7 mil
Thickness (Metric) 0.12 mm
Length (English) 9.6 in.
Length (Metric) 245 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.