missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microcystin-LR Antibody (MC10E7) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89027JF635
This item is not returnable.
View return policy
Description
Microcystin-LR Monoclonal antibody specifically detects Microcystin-LR in Nonspecies specific samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Microcystin-LR | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Non-species specific | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| MC10E7 | |
| 50mM Sodium Borate | |
| Microcystin-LR linked via N-methyl-dehydroalanine to cationized ovalbumin. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction