missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microcystin-LR Antibody (MC10E7), Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Microcystin-LR Monoclonal antibody specifically detects Microcystin-LR in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Microcystin-LR |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MC10E7 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Microcystin-LR linked via N-methyl-dehydroalanine to cationized ovalbumin. |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?