missing translation for 'onlineSavingsMsg'
Learn More

Microcystin-LR Antibody (MC10E7), DyLight 550, Novus Biologicals™

Product Code. 18415366 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415366 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415366 Supplier Novus Biologicals Supplier No. NBP289027R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Microcystin-LR Monoclonal antibody specifically detects Microcystin-LR in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Microcystin-LR
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MC10E7
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Microcystin-LR linked via N-methyl-dehydroalanine to cationized ovalbumin.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.