missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Micrococcal nuclease (S. aureus)

Product Code. 16151820
Click to view available options
Quantity:
15,000 U
Unit Size:
15000 units
This item is not returnable. View return policy

Product Code. 16151820

Brand: Abnova™ P5291.15000Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 16.8kDa
Name Micrococcal nuclease (S. aureus)
Preparation Method Native protein chromatographically purified from Staphylococcus aureus (Strain Foggi)
Quantity 15,000 U
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name MICROCOCCAL
Assays Micrococcal Nuclease Assay; Elucidation of nucleic acid and oligonucleotide sequence studies.
Biological Activity ≥6,000 units per mg protein. One Unit corresponds to a change in optical density of 1.0 at 260nm at 37°C, pH 8.0, using DNA as the substrate.
Species Staphylococcus species
Protein Tag None
Expression System Native protein chromatographically purified from Staphylococcus aureus (Strain Foggi)
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.