missing translation for 'onlineSavingsMsg'
Learn More

MicroChem™ Worksheets

Product Code. 12826955
Click to view available options
Grade:
KS 3
KS 4
KS 5
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 12826955

Brand: Edulab S004/B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Grade KS 4
Format Sheet
Product Type MicroChem Worksheets
Includes Risk assessment, List of equipment and chemicals required, Relevant specifications covered for AQA, EDEXCEL, OCR and QCA
Quantity 1/Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.