Learn More
Emma Safety Footwear Micro Safety Shoes
Product Code. 15958451
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15958451
Brand: Emma Safety Footwear 50054040
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Unique microfiber upper is fully washable. SmartecAir™ provides fresh dry feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors used in high-tech and cleanroom environments.
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Elastic
Color White
Design Micro
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 40
Toe Material Steel