Learn More
BOLA Micro Reaction Vessels
Product Code. p-200109995
Click to view available options
For Use With (Equipment):
connections
threads
Unit Size:
Each
Product Code. 10737872
Brand: BOLA B31840
Micro Reaction Vessels Made of PFA. With a PTFE lid to be screwed off.
Vessel made of translucent, non-porous PFA, screw cap made of PTFE with threaded necks for connection
Specifications
For Use With (Equipment) connections
Autoclavable Autoclavable