Learn More
Fisherbrand™ Micro Insert for well plates, glass
For complex UHPLC, LC and GC applications.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Clear
- Glass
- Plates meet ANSI and SBS standards
- Different sealing options
- Chromatography tested
- No leeching due to high-end material
Specifications
Clear | |
Glass | |
1000 μL | |
Round |