Learn More
Specac™ Micro Compression Cell Windows NaCl
Product Code. 15280078
Encompass_Preferred
Click to view available options
Type:
Micro Compression Cell Windows NaCl
Product Code. 15280078
Brand: Specac™ GS09070
This cell is designed for soft solid of semi liquid sample analysis by FTIR transmission or infrared microscopy. This cell is fitted with NaCl windows and allows for samples to be compressed and flattened between two non-rotating transmission windows for analysis. Clear aperture of 7.0mm and can be used with any FTIR instrument.
Specifications
Type Micro Compression Cell Windows NaCl
Description Clear aperture of 7.0 mm and can be used with any FTIR instrument