Learn More
Specac™ Micro Compression Cell Windows KBr
Product Code. 15290078
Encompass_Preferred
Change view
Click to view available options
Type:
Micro Compression Cell Windows KBr
Product Code. Type
15290078 Micro Compression Cell Windows KBr
1 options
Product Code. 15290078 Supplier Specac™ Supplier No. GS09071
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This cell is designed for soft solid of semi liquid sample analysis by FTIR transmission or infrared microscopy. This cell is fitted with KBr windows and allows for samples to be compressed and flattened between two non-rotating transmission windows for analysis. Clear aperture of 7.0mm and can be used with any FTIR instrument.
Specifications
Type Micro Compression Cell Windows KBr
Description Clear aperture of 7.0 mm and can be used with any FTIR instrument