Learn More
Specac™ Micro Compression Cell Windows CaF2
Product Code. 15200088
Encompass_Preferred
Change view
Click to view available options
Type:
Micro Compression Cell Windows CaF2
Product Code. Type
15200088 Micro Compression Cell Windows CaF2
1 options
Product Code. 15200088 Supplier Specac™ Supplier No. GS09072
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This cell is designed for soft solid of semi liquid sample analysis by FTIR transmission or infrared microscopy. This cell is fitted with CaF2 windows and allows for samples to be compressed and flattened between two non-rotating transmission windows for analysis. Clear aperture of 7.0mm and can be used with any FTIR instrument.
Specifications
Type Micro Compression Cell Windows CaF2
Description Clear aperture of 7.0 mm and can be used with any FTIR instrument