Learn More
Specac™ Micro Compression Cell Windows BaF2
Product Code. 15210088
Encompass_Preferred
Click to view available options
Type:
Micro Compression Cell Windows BaF2
Product Code. 15210088
Brand: Specac™ GS09073
This cell is designed for soft solid of semi liquid sample analysis by FTIR transmission or infrared microscopy. This cell is fitted with BaF2 windows and allows for samples to be compressed and flattened between two non-rotating transmission windows for analysis. Clear aperture of 7.0mm and can be used with any FTIR instrument.
Specifications
Type Micro Compression Cell Windows BaF2
Description Clear aperture of 7.0 mm and can be used with any FTIR instrument