missing translation for 'onlineSavingsMsg'
Learn More

Miconazole Nitrate, Mikromol™

Product Code. 17971516
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971516 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971516 Supplier Mikromol Supplier No. MM0281.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Miconazole Nitrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Miconazole Nitrate
CAS 22832-87-7
Analyte or Component Names Miconazole Nitrate
Shipping Condition Room Temperature
Molecular Formula C18 H14 Cl4 N2 O . H N O3
Certifications/Compliance ISO 17034
Synonym 1-[(2RS)-2-[(2,4-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate, Isoconazole Nitrate Imp. C (EP) as Nitrate, 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole nitrate (1:1), 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole mononitrate, Aflorix, Albistat, Andergin, Antifungal Cream, Brentan, Conoderm, Conofite, Daktacort, Daktar, Daktarin, Daktarin talc, Deralbine, Dermonistat, Ecobi, Epi-Monistat, Florid, Fungiderm, Fungisdin, Gyno-Daktarin, Gyno-Monistat, Hi-Pick, Loptrimin AF Jock-Itch Powder Aerosol, Lotrimin AF Powder, Lotrimin AF Powder Aerosol, Lotrimin AF Spray Liquid, Micatin, Miconal, Miconaz, Miconazole nitrate, Micotef, Monistat, Monistat Cream and Suppositories, Monistat-Derm, NSC 169434, Priconazole, Prilagin, R 14889, Vodol, Zeasorb AF, Zimycan, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-, mononitrate (9CI), Imidazole, 1-[2,4-dichloro-beta-[(2,4-dichlorobenzyl)oxy]phenethyl]-, mononitrate (8CI), (+/-)-Miconazole nitrate, Dactrine, [2,4-Dichloro-beta-(2,4-dichlorobenzyloxy)phenethyl]imidazole nitrate, Isoconazole Nitrate Impurity C as Nitrate, Isoconazole Impurity C as Nitrate
InChI Formula InChI=1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)
SMILES O[N+](=O)[O-].Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid
Molecular Weight (g/mol) 479.14
Quantity 250 mg
Formula Weight 476.9817 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.