missing translation for 'onlineSavingsMsg'
Learn More

Miconazole nitrate, MedChemExpress

Product Code. 16480938
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480938 5 g 5g
16402189 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480938 Supplier Medchem Express Supplier No. HYB0454A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Miconazole nitrate (R18134 nitrate) is an imidazole antifungal agent. Miconazole nitrate also has antibacterial effects.

Miconazole nitrate (R18134 nitrate) is an imidazole antifungal agent. Miconazole nitrate also has antibacterial effects.

Specifications

Chemical Name or Material Miconazole nitrate
Physical Form Solid
CAS 22832-87-7
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C18H15Cl4N3O4
For Use With (Application) COVID-19-immunoregulation
Synonym R18134 nitrate
Solubility Information DMSO : 50 mg/mL (104.35 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [O-][N+](O)=O.ClC1=C(C=CC(Cl)=C1)C(OCC(C=CC(Cl)=C2)=C2Cl)CN3C=CN=C3
Molecular Weight (g/mol) 479.14
Formula Weight 479.14
Percent Purity 99.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.