Learn More
(+/-)-Miconazole nitrate, 98+%, Thermo Scientific™
Inhibits cytochrome P450-dependent 14a-demethylase
£79.70 - £199.00
Chemical Identifiers
| CAS | 22832-87-7 |
|---|---|
| Molecular Formula | C18H15Cl4N3O4 |
| Molecular Weight (g/mol) | 479.14 |
| MDL Number | MFCD00058161 |
| InChI Key | MCCACAIVAXEFAL-UHFFFAOYNA-N |
| Synonym | miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar |
| PubChem CID | 68553 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid |
| SMILES | O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15493499
|
Thermo Scientific Alfa Aesar
J60796.06 |
5g |
£79.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15403509
|
Thermo Scientific Alfa Aesar
J60796.14 |
25g |
£199.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 22832-87-7 | |
| 479.14 | |
| MCCACAIVAXEFAL-UHFFFAOYNA-N | |
| 68553 | |
| O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1 |
| C18H15Cl4N3O4 | |
| MFCD00058161 | |
| miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar | |
| 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid |
Specifications
| 22832-87-7 | |
| MFCD00058161 | |
| miconazole nitrate, albistat, andergin, aflorix, conofite, florid, micatin, gyno-monistat, epi-monistat, gyno-daktar | |
| MCCACAIVAXEFAL-UHFFFAOYNA-N | |
| 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole; nitric acid | |
| 68553 | |
| ≥98% |
| C18H15Cl4N3O4 | |
| 14,6178 | |
| Slightly soluble in water; Soluble in propylene glycol or pyridine. | |
| O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1 | |
| 479.14 | |
| 479.15 | |
| (+/-)-Miconazole nitrate |
Safety and Handling
GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
missing translation for 'einecsNumber' : 245-256-6
missing translation for 'rtecsNumber' : NI4771000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.