missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miconazole, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Miconazole
Specifications
Specifications
| Chemical Name or Material | Miconazole |
| CAS | 22916-47-8 |
| Analyte or Component Names | Miconazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H14 Cl4 N2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Miconazole, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-, Daktanol, Daktarin IV, Florid-F, Lauriad, MJR 1762, Monistat IV, NSC 170986, R 18134, Zimybase, Imidazole, 1-[2,4-dichloro-beta-[(2,4-dichlorobenzyl)oxy]phenethyl]- (8CI), 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole, (+/-)-Miconazole, Loramyc, Isoconazole Impurity D |
| InChI Formula | InChI=1S/C18H14Cl4N2O/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22/h1-8,11,18H,9-10H2 |
| SMILES | Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction