missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICL/CLEC12A Antibody (2439B) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB29461JF525
This item is not returnable.
View return policy
Description
MICL/CLEC12A Monoclonal antibody specifically detects MICL/CLEC12A in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,CyTOF
Specifications
| MICL/CLEC12A | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
| Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A, Thr67-Ala265, Accession # Q5QGZ9 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 2439B | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 160364 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction