missing translation for 'onlineSavingsMsg'
Learn More

Micheliolide, MedChemExpress

Product Code. 16469128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469128 10 mg 10mg
16421538 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469128 Supplier Medchem Express Supplier No. HYN0847
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Micheliolide could effectively attenuate the high glucose-stimulated activation of NF-κB, the degradation of IκBα, and the expression of MCP-1, TGF-β1 and FN in rat mesangial cells (MCs).

Micheliolide could effectively attenuate the high glucose-stimulated activation of NF-κB, the degradation of IκBα, and the expression of MCP-1, TGF-β1 and FN in rat mesangial cells (MCs).

Specifications

Chemical Name or Material Micheliolide
CAS 68370-47-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H20O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (402.71 mM; Need ultrasonic)
SMILES O=C(O[C@@]1([H])[C@@]2([H])CCC(C)=C3CC[C@@](C)(O)[C@@]31[H])C2=C
Molecular Weight (g/mol) 248.32
Formula Weight 248.32
Percent Purity 99.17%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.