missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICB Mouse anti-Human, Clone: B-D55, Novus Biologicals™
Mouse Monoclonal Antibody
£305.00 - £571.00
Specifications
| Antigen | MICB |
|---|---|
| Clone | B-D55 |
| Concentration | 1 mg/ml |
| Dilution | ELISA |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18396235
|
Novus Biologicals
NBP3-18103-200UL |
200 μg |
£305.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18376826
|
Novus Biologicals
NBP3-18103-500UL |
500 μg |
£571.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for ELISAEspecificaciones
| MICB | |
| 1 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins | |
| PBS. Sterile-filtered through 0.22 um. | |
| 4277 | |
| IgG1 κ | |
| Affinity purified |
| B-D55 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
| Recombinant human MICB. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto