missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICB Antibody (236511) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1599JF669
This item is not returnable.
View return policy
Description
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for Flow Cytometry,Western Blot,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF,Gene Knock-Out
Specifications
| MICB | |
| Monoclonal | |
| Janelia Fluor 669 | |
| MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
| Mouse myeloma cell line NS0-derived recombinant human MICB, Ala23-Gly298, Accession # CAI18747 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF, Gene Knock-Out | |
| 236511 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 4277 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction