missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICALL1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) H00085377-B01P
This item is not returnable.
View return policy
Description
MICALL1 Polyclonal antibody specifically detects MICALL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| MICALL1 | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:400 | |
| Q8N3F8 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.4) | |
| DKFZp686M2226, KIAA1668, MICAL-L1, MICAL-like 1, MICAL-like protein 1 | |
| MICAL-L1 (AAI48805.1, 1 a.a. - 863 a.a.) full-length human protein. MAGPRGALLAWCRRQCEGYRGVEIRDLSSSFRDGLAFCAILHRHRPDLLDFDSLSKDNVFENNRLAFEVAEKELGIPALLDPNDMVSMSVPDCLSIMTYVSQYYNHFCSPGQAGVSPPRKGLAPCSPPSVAPTPVEPEDVAQGEELSSGSLSEQGTGQTPSSTCAACQQHVHLVQRYLADGRLYHRHCFRCRRCSSTLLPGAYENGPEEGTFVCAEHCARLGPGTRSGTRPGPFSQPKQQHQQQLAEDAKDVPGGGPSSSAPAGAEADGPKASPEARPQIPTKPRVPGKLQELASPPAGRPTPAPRKASESTTPAPPTPRPRSSLQQENLVEQAGSSSLVNGRLHELPVPKPRGTPKPSEGTPAPRKDPPWITLVQAEPKKKPAPLPPSSSPGPPSQDSRQVENGGTEEVAQPSPTASLESKPYNPFEEEEEDKEEEAPAAPSLATSPALGHPESTPKSLHPWYGITPTSSPKTKKRPAPRAPSASPLALHASRLSHSEPPSATPSPALSVESLSSESASQTAGAELLEPPAVPKSSSEPAVHAPGTPGNPVSLSTNSSLASSGELVEPRVEQMPQASPGLAPRTRGSSGPQPAKPCSGATPTPLLLVGDRSPVPSPGSSSPQLQVKSSCKENPFNRKPSPAASPATKKATKGSKPVRPPAPGHGFPLIKRKVQADQYIPEEDIHGEMDTIERRLDALEHRGVLLEEKLRGGLNEGREDDMLVDWFKLIHEKHLLVRRESELIYVFKQQNLEQRQADVEYELRCLLNKPEKDWTEEDRAREKVLMQELVTLIEQRNAIINCLDEDRQREEEEDKMLEAMIKKKEFQREAEPEGKKKGKFKTMKMLKLLGNKRDAKSKSPRDKS | |
| 0.05 mg | |
| Signal Transduction | |
| 85377 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction