Learn More
MICAL3 Antibody (3A6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00057553-M08
Product Code. 18498038
  • £463.00 / 0.10mg
Description
MICAL3 Monoclonal antibody specifically detects MICAL3 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MICAL3
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
microtubule associated monoxygenase, calponin and LIM domain containing 3
MICAL3 (XP_032996.2, 251 a.a. ∽ 340 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DDQHWSDSPSDADRELRLPCPAEGEAELELRVSEDEEKLPASPKHQERGPSQATSPIRSPQESALLFIPVHSPSTEGPQLPPVPAATQEK
0.1 mg
Primary
Human
Purified
Western Blot, ELISA, Sandwich ELISA
3A6
Western Blot, ELISA, Sandwich ELISA
XP_032996.2
Mouse
IgG purified
RUO
57553
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG2a κ