missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICAL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35565-100ul
This item is not returnable.
View return policy
Description
MICAL1 Polyclonal antibody specifically detects MICAL1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MICAL1 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| CasL interacting molecule, DKFZp434B1517, FLJ11937, FLJ21739, MICALMolecule interacting with CasL protein 1, microtubule associated monoxygenase, calponin and LIM domain containing 1, NEDD9 interacting protein with calponin homology and LIM domains, NEDD9-interacting protein with calponin homology and LIM domains, NICALMICAL-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 718-1067 of human MICAL1 (NP_073602.3).,, Sequence:, HRSCFRCHTCEATLWPGGYEQHPGDGHFYCLQHLPQTDHKAEGSDRGPESPELPTPSENSMPPGLSTPTASQEGAGPVPDPSQPTRRQIRLSSPERQRLSSLNLTPDPEMEPPPKPPRSCSALARHALESSFVGWGLPVQSPQALVAMEKEEKESPFSSEEEEEDVPLDSDVEQALQTFAKTSGTMNNYPTWRRTLLRRAKEEEMKRFCKAQTIQRRLNEIEAALRELEAEGVKLELALRRQSSSPEQQKKLWVGQLLQLVDKKNSLVAEEAELMITVQELNLEEKQWQLDQELRGYMNREENLKTAADRQAEDQVLRKLVDLVNQRDALIRFQEERRLSELALGTGAQG | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64780 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction