missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micafungin sodium, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Micafungin sodium (FK 463 sodium) is an antifungal agent which inhibits 1, 3-beta-D-glucan synthesis.
Specifications
Specifications
| Chemical Name or Material | Micafungin sodium |
| Physical Form | Solid |
| CAS | 208538-73-2 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C56H70N9NaO23S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | FK 463 sodium |
| Solubility Information | H2O : 100 mg/mL (77.38 mM; Need ultrasonic) ∣DMSO : ≥ 32 mg/mL (24.76 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction