Learn More
Invitrogen™ MICA Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ BMS2302
300.68 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.40000kg
Description
The Human MICA ELISA quantitates Hu MICA in human serum, plasma, cell lysates, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MICA.
Principle of the method
The Human MICA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Q29983 | |
1.09 pg/ml | |
ELISA Kit | |
Human | |
100507436 | |
6.6% | |
Pre-coated 96-well Plate, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromagen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Serum, 50 μL; Plasma, 50 μL; Cell Lysate,50 μL; Supernatant, 50 μL | |
Human | |
4 hr. |
1.6-100 pg/ml | |
Biotin | |
Cell Lysates, Plasma, Serum, Supernatant | |
Colorimetric Microplate Reader | |
MICA | |
5.1% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |