missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICA Antibody (159227) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1300Y
This item is not returnable.
View return policy
Description
MICA Monoclonal antibody specifically detects MICA in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, Gene Knock-Out
Specifications
| MICA | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF, Gene Knock-Out | |
| 159227 | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready, Knockout Validated | |
| FLJ60820, MGC111087, PERB11.1 | |
| BaF3 mouse pro-B cell line transfected with human MICA | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 100507436 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction