Learn More
MIC-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560428
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CLLLSLESFY PPAHQLSLDM LKRLSTANDE IVEVLLSKHQ VLAALRFIRG IGGHDNISAR KFLDAAKQTE DNMLFYTIFR FFEQRNQRLR GSPNFTPGE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
Colon cancer-associated protein Mic1.Specifications
MIC-1 | |
Polyclonal | |
Unconjugated | |
Rmc1 | |
2400010D15Rik; 3110002H16Rik; C18orf8; chromosome 18 open reading frame 8; colon cancer-associated protein Mic1; colon cancer-associated protein Mic1 homolog; HsT2591; macrophage inhibitory cytokine 1; Mic1; Mic-1; Regulator of MON1-CCZ1 complex; RGD1311805; RIKEN cDNA 3110002H16 gene; RMC1; similar to RIKEN cDNA 2400010D15; uncharacterized protein C18orf8; uncharacterized protein C18orf8 homolog; WD repeat-containing protein 98; WDR98 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29919 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DM3 | |
Rmc1 | |
Recombinant protein corresponding to Human MIC-1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only