missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIC-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571579
This item is not returnable.
View return policy
Description
MIC-1 Polyclonal Antibody for Western Blot, ICC/IF, Flow
Colon cancer-associated protein Mic1.
Specifications
| MIC-1 | |
| Polyclonal | |
| Unconjugated | |
| Rmc1 | |
| 2400010D15Rik; 3110002H16Rik; C18orf8; chromosome 18 open reading frame 8; colon cancer-associated protein Mic1; colon cancer-associated protein Mic1 homolog; HsT2591; macrophage inhibitory cytokine 1; Mic1; Mic-1; Regulator of MON1-CCZ1 complex; RGD1311805; RIKEN cDNA 3110002H16 gene; RMC1; similar to RIKEN cDNA 2400010D15; uncharacterized protein C18orf8; uncharacterized protein C18orf8 homolog; WD repeat-containing protein 98; WDR98 | |
| Rabbit | |
| Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 29919 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q96DM3 | |
| Rmc1 | |
| KLH conjugated synthetic peptide between 34-66 amino acids from the N-terminal region of human MIC1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction