missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mibefradil dihydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Mibefradil dihydrochloride (Ro 40-5967 dihydrochloride) is a calcium channel blocker with moderate selectivity for T-type Ca2+ channels (IC50s of 2.7 μM and 18.6 μM for T-type and L-type currents, respectively).
Specifications
Specifications
| Chemical Name or Material | Mibefradil dihydrochloride |
| CAS | 116666-63-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H40Cl2FN3O3 |
| Quantity | 5 mg |
| Synonym | Ro 40-5967 dihydrochloride |
| Solubility Information | H2O : ≥ 125 mg/mL (219.86 mM) |
| SMILES | O=C(O[C@@]1(CCN(CCCC2=NC3=CC=CC=C3N2)C)[C@@H](C(C)C)C4=C(C=C(F)C=C4)CC1)COC.[H]Cl.[H]Cl |
| Molecular Weight (g/mol) | 568.55 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction