missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572810
This item is not returnable.
View return policy
Description
MIB1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
MIB is an E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. This protein positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors. MIB probably mediates ubiquitination and subsequent proteasomal degradation of DAPK1, thereby antagonizing anti-apoptotic effects of DAPK1 to promote TNF-induced apoptosis.
Specifications
| MIB1 | |
| Polyclonal | |
| Unconjugated | |
| MIB1 | |
| DAPK-interacting protein 1; DAPK-interacting protein-1; DIP1; DIP-1; E3 ubiquitin-protein ligase MIB1; E430019M12Rik; KIAA1323; LVNC7; MIB; MIB1; Mind bomb homolog 1; mind bomb-1; mindbomb E3 ubiquitin protein ligase 1; mindbomb homolog 1; mindbomb homolog 1 (Drosophila); mKIAA1323; RING-type E3 ubiquitin transferase MIB1; skeletrophin; ubiquitin ligase mind bomb; zinc finger ZZ type with ankyrin repeat domain protein 2; ZZANK2; ZZZ6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 225164, 307594, 57534 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q80SY4, Q86YT6 | |
| MIB1 | |
| MIB1 antibody for a 19 amino acid peptide near the carboxy terminus of human MIB1. The immunogen is within amino acids 920 - 970 of MIB1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction