missing translation for 'onlineSavingsMsg'
Learn More

Mianserin hydrochloride, MedChemExpress

Product Code. 16451396
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16451396

Brand: Medchem Express HYB0188A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mianserin hydrochloride (Org GB 94) is a H1 receptor inverse agonist and is a psychoactive agent of the tetracyclic antidepressant.

Mianserin hydrochloride (Org GB 94) is a H1 receptor inverse agonist and is a psychoactive agent of the tetracyclic antidepressant.

Specifications

Chemical Name or Material Mianserin hydrochloride
CAS 21535-47-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H21ClN2
Quantity 200 mg
Synonym Org GB 94
Solubility Information DMSO : 50 mg/mL (166.21 mM; Need ultrasonic) ∣H2O : 25 mg/mL (83.10 mM; ultrasonic and warming and heat to 60°C)
SMILES CN(CC1)CC2N1C3=CC=CC=C3CC4=CC=CC=C24.Cl
Molecular Weight (g/mol) 300.83
Formula Weight 300.83
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.