missing translation for 'onlineSavingsMsg'
Learn More

Mianserin hydrochloride, MedChemExpress

Product Code. 16462056
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462056 500 mg 500mg
16458166 100 mg 100mg
16451396 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16462056 Supplier Medchem Express Supplier No. HYB0188A
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mianserin hydrochloride (Org GB 94) is a H1 receptor inverse agonist and is a psychoactive agent of the tetracyclic antidepressant.

Mianserin hydrochloride (Org GB 94) is a H1 receptor inverse agonist and is a psychoactive agent of the tetracyclic antidepressant.

Specifications

Chemical Name or Material Mianserin hydrochloride
CAS 21535-47-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H21ClN2
Quantity 500 mg
Synonym Org GB 94
Solubility Information DMSO : 50 mg/mL (166.21 mM; Need ultrasonic) ∣H2O : 25 mg/mL (83.10 mM; ultrasonic and warming and heat to 60°C)
SMILES CN(CC1)CC2N1C3=CC=CC=C3CC4=CC=CC=C24.Cl
Molecular Weight (g/mol) 300.83
Formula Weight 300.83
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.