missing translation for 'onlineSavingsMsg'
Learn More

Mianserin.HCl, 1mg/ml in Methanol (as free base), Lipomed

Product Code. 30459760
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459760 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459760 Supplier Lipomed Supplier No. LPMMSR1438HC1LM
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Mianserin.HCl, 1mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mianserin Hydrochloride
CAS 21535-47-7
Analyte or Component Names Mianserin Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H20 N2 . Cl H
Concentration 1 mg/mL
Synonym Mianserin Hydrochloride, Dibenzo[c,f]pyrazino[1,2-a]azepine, 1,2,3,4,10,14 b-hexahydro-2-methyl-, hydrochloride (1:1), Dibenzo[c,f]pyrazino[1,2-a]azepine, 1,2,3,4,10,14 b-hexahydro-2-methyl-, monohydrochloride, Athymil, Bolvidon, GB 94, Lantanon, Mianserin hydrochloride, Mianserine hydrochloride, NSC 292267, Norval, Org GB 94, Tetramide, Toluon, Tolvin, Tolvon
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.CN1CCN2C(C1)c3ccccc3Cc4ccccc24
IUPAC Name 1,2,3,4,10,14 b-Hexahydro-2-methyldibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 300.83
Quantity 1 mL
Formula Weight 300.1393 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.