missing translation for 'onlineSavingsMsg'
Learn More

(±)-Mianserin-d3 HCl (methyl-d3), TRC

Product Code. 30478567
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.5mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478567 0.5 mg 0.5mg
30467194 5 mg 5mg
30470764 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478567 Supplier Toronto Research Chemicals Supplier No. TRCM3415010.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219804-97-3
Molecular Formula C18 D3 H17 N2 . Cl H
Molecular Weight (g/mol) 303.8442
Synonym (±)-Mianserin-d3 HCl (methyl-d3)
IUPAC Name 5-(trideuteriomethyl)-2,5-diazatetracyclo[13.4.0.0^{2,7}.0^{8,13}]nonadeca-1(19),8(13),9,11,15,17-hexaene;hydrochloride
SMILES Cl.[2H]C([2H])([2H])N1CCN2C(C1)c3ccccc3Cc4ccccc24

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C18H20N2.ClH/c1-19-10-11-20-17-9-5-3-7-15(17)12-14-6-2-4-8-16(14)18(20)13-19;/h2-9,18H,10-13H2,1H3;1H/i1D3;
Formula Weight 303.1582
Chemical Name or Material (±)-Mianserin-d3 Hydrochloride (methyl-d3)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.