missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17481-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and contains a predicted leucine zipper motif typical of many transcription factors.Specifications
| MIA3 | |
| Polyclonal | |
| Unconjugated | |
| MIA3 | |
| ARNT, C219 reactive peptide, D320, KIAA0268, MIA3, TANGO, TANGO1, UNQ6077 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 338366, 375056, 683007 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5JRA6, Q8BI84 | |
| Mia3 | |
| MIA3 Fusion Protein Ag11514 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction