missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106940
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MIA3.
Plays a role in the transport of cargos that are too large to fit into COPII-coated vesicles and require specific mechanisms to be incorporated into membrane-bound carriers and exported from the endoplasmic reticulum. This protein is required for collagen VII (COL7A1) secretion by loading COL7A1 into transport carriers. It may participate in cargo loading of COL7A1 at endoplasmic reticulum exit sites by binding to COPII coat subunits Sec23/24 and guiding SH3-bound COL7A1 into a growing carrier. Does not play a role in global protein secretion and is apparently specific to COL7A1 cargo loading. However, it may participate in secretion of other proteins in cells that do not secrete COL7A1. It is also specifically required for the secretion of lipoproteins by participating in their export from the endoplasmic reticulum (PubMed:27138255, PubMed:19269366). Required for correct assembly of COPII coat components at endoplasmic reticulum exit sites (ERES) and for the localization of SEC16A and membrane-bound ER-resident complexes consisting of MIA2 and PREB/SEC12 to ERES (PubMed:28442536). [UniProt].
Specifications
| MIA3 | |
| Polyclonal | |
| Unconjugated | |
| MIA3 | |
| 9130229H14Rik; A930039G15Rik; ARNT; AU022327; B230399H06Rik; C219-reactive peptide; C80126; D320; Gm1525; Kiaa0268; melanoma inhibitory activity 3; melanoma inhibitory activity family member 3; melanoma inhibitory activity family, member 3; melanoma inhibitory activity protein 3; MIA family member 3, ER export factor; Mia3; RP11-378J18.4; Tango; TANGO1; transport and Golgi organization protein 1; Transport and Golgi organization protein 1 homolog; UNQ6077; UNQ6077/PRO20088 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 338366, 375056, 683007 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5JRA6, Q8BI84 | |
| MIA3 | |
| A synthesized peptide derived from human MIA3(Accession Q5JRA6), corresponding to amino acid residues P1844-S1894. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction