missing translation for 'onlineSavingsMsg'
Learn More

PeproTech MIA Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Product Code. 17835263
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
50 μg
500 μg
Unit Size:
1mg
25µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17835263 25 μg 25µg
17805393 50 μg 50µg
17885763 2 x 500 μg 1mg
30054720 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17835263 Supplier PeproTech Supplier No. 500P243BT25UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MGPMPKLADR KLCADQECSH PISMAVALQD YMAPDCRFLT IHRGQVVYVF SKLKGRGRLF WGGSVQGDYY GDLAARLGYF PSSIVREDQT LKPGKVDVKT DKWDFYCQ. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MIA. Anti-Human MIA-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hMIA by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human MIA (500-P243) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hMIA. Western Blot: To detect hMIA by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hMIA is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P243BT-1MG will be provided as 2 x 500 μg

Tumorigenesis is a process that is mediated by a network of membrane, cytosolic and nuclear associated factors, which regulate proliferation and cell-matrix interaction through signaling cascades. The phenotype of malignant melanomas in vivo depends on the global expression of stimulatory or inhibitory factors generated in both the tumors cells and their environment. One example, Melanoma inhibitory activity (cartilage-derived retinoic acid-sensitive protein (CD-RAP), MIA) is a Src homology 3 (SH3)-like domain containing protein that is secreted from chondrocytes and malignant melanoma cells. MIA is translated as a 131-amino acid pro-form and processed into a mature 107-amino acid protein after cleavage of a secretion signal. MIA is expressed during chondrogenesis and in mature chondrocytes, suggesting that MIA is necessary for normal cartilage cell phenotype. MIA mRNA is present in carcinomas of the colon, ovary, kidney, and head/neck, and may represent a marker to monitor melanomic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene MIA
Gene Accession No. Q16674
Gene Alias cartilage derived retinoic acid sensitive protein; cartilage-derived retinoic acid-sensitive protein; Cdrap; CD-RAP; melanoma inhibitory activity; melanoma inhibitory activity 1; melanoma inhibitory activity protein; melanoma inhibitory activity/condrocyte-derived retinoic acid sensitive protein homolog (MIA/CD-RAP); melanoma-derived growth regulatory protein; melanoma-inhibitory-activity; MIA; Mia1
Gene Symbols MIA
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human MIA.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8190
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.