missing translation for 'onlineSavingsMsg'
Learn More

PeproTech MIA Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P243BT-50UG

 View more versions of this product

Product Code. 17805393

  • £228.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MGPMPKLADR KLCADQECSH PISMAVALQD YMAPDCRFLT IHRGQVVYVF SKLKGRGRLF WGGSVQGDYY GDLAARLGYF PSSIVREDQT LKPGKVDVKT DKWDFYCQ. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MIA. Anti-Human MIA-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hMIA by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human MIA (500-P243) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hMIA. Western Blot: To detect hMIA by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hMIA is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P243BT-1MG will be provided as 2 x 500 μg

Tumorigenesis is a process that is mediated by a network of membrane, cytosolic and nuclear associated factors, which regulate proliferation and cell-matrix interaction through signaling cascades. The phenotype of malignant melanomas in vivo depends on the global expression of stimulatory or inhibitory factors generated in both the tumors cells and their environment. One example, Melanoma inhibitory activity (cartilage-derived retinoic acid-sensitive protein (CD-RAP), MIA) is a Src homology 3 (SH3)-like domain containing protein that is secreted from chondrocytes and malignant melanoma cells. MIA is translated as a 131-amino acid pro-form and processed into a mature 107-amino acid protein after cleavage of a secretion signal. MIA is expressed during chondrogenesis and in mature chondrocytes, suggesting that MIA is necessary for normal cartilage cell phenotype. MIA mRNA is present in carcinomas of the colon, ovary, kidney, and head/neck, and may represent a marker to monitor melanomic activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MIA
Polyclonal
Biotin
MIA
cartilage derived retinoic acid sensitive protein; cartilage-derived retinoic acid-sensitive protein; Cdrap; CD-RAP; melanoma inhibitory activity; melanoma inhibitory activity 1; melanoma inhibitory activity protein; melanoma inhibitory activity/condrocyte-derived retinoic acid sensitive protein homolog (MIA/CD-RAP); melanoma-derived growth regulatory protein; melanoma-inhibitory-activity; MIA; Mia1
Rabbit
Antigen affinity chromatography
RUO
8190
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
Q16674
MIA
E.coli-derived Recombinant Human MIA.
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.