missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MI 2 (MALT1 inhibitor)
GREENER_CHOICE

Product Code. 30001668
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001668 50 mg 50mg
30001654 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001668 Supplier Tocris Bioscience™ Supplier No. 4848/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MALT1 inhibitor

MALT1 inhibitor (IC50 = 5.84 μM). Binds directly to MALT1 and irreversibly suppresses protease function. Decreases NF-κB activity induced by MALT1. Inhibits cell proliferation and MALT1-mediated cleavage activity. Suppresses human TMD8 and HBL-1 activated B cell-like diffuse large B cell lymphoma (ABC-DLBCL) tumor xenografts in mice and primary human ABC-DLBCLs ex vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1047953-91-2
Molecular Formula C19H17Cl3N4O3

Specifications

Quantity 50 mg
Formula Weight 455.72
Purity >98%
Molecular Weight (g/mol) 455.72
Chemical Name or Material MI 2 (MALT1 inhibitor)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.