missing translation for 'onlineSavingsMsg'
Learn More

MHY1485, MedChemExpress

Product Code. 16484487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484487

Brand: Medchem Express HYB0795

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MHY1485 is a potent cell-permeable mTOR activator that targets the ATP domain of mTOR. MHY1485 inhibits autophagy by suppression of fusion between autophagosomes and lysosomes.

MHY1485 is a potent cell-permeable mTOR activator that targets the ATP domain of mTOR. MHY1485 inhibits autophagy by suppression of fusion between autophagosomes and lysosomes.

Specifications

Chemical Name or Material MHY1485
Physical Form Solid
CAS 326914-06-1
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H21N7O4
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 7.69 mg/mL (19.85 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.58 mM; Need ultrasonic)
SMILES O=[N+](C1=CC=C(NC2=NC(N3CCOCC3)=NC(N4CCOCC4)=N2)C=C1)[O-]
Molecular Weight (g/mol) 387.39
Formula Weight 387.39
Percent Purity 98.29%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.