missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHP, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
Description
MHP (Methyl caprooyl tyrosinate) is an activator of sphingosine kinase (SPHK1), and significantly stimulates CAMP mRNA and protein production. MHP (Methyl caprooyl tyrosinate) enhances antimicrobial defense and innate immunity.
Specifications
Specifications
| Chemical Name or Material | MHP |
| CAS | 1104874-94-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H23NO4 |
| Quantity | 10 mg |
| Synonym | Methyl caprooyl tyrosinate |
| Solubility Information | DMSO : ≥ 100 mg/mL (340.88 mM) ∣H2O : 1 mg/mL (3.41 mM; Need ultrasonic) |
| SMILES | OC(C=C1)=CC=C1C[C@@H](C(OC)=O)NC(CCCCC)=O |
| Molecular Weight (g/mol) | 293.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction