Learn More
Invitrogen™ MHC I Free Chain Monoclonal Antibody (A4), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£168.00 - £3115.00
Specifications
| Antigen | MHC I Free Chain |
|---|---|
| Clone | A4 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15258277
![]() |
Invitrogen™
14-9958-82 |
100 μg |
£168.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17140842
|
Invitrogen™
14-9958-37 |
2 mg |
£1376.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17150842
|
Affymetrix eBioscience
14-9958-95 |
10 mg | |||||||
Description
Description: The A4 monoclonal antibody reacts with the class I human leukocyte antigen (HLA) complex in the absence of beta2 microglobulin. There are three class I alpha-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with beta2 microglobulin (beta2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells. Applications Reported: This A4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of paraffin embedded tissue sections. Applications Tested: This A4 antibody has been tested by flow cytometric analysis of 3-day PHA-activated normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
There are three class I alpha-chain genes in humans, called HLA-A, -B and -C. Normally, HLA class I proteins are associated non-covalently with B2 microglobulin (B2M). The function of the HLA class I complex is to present peptides derived from intracellular pathogens, including viral pathogens, on the cell surface to antigen specific CD8+ cytotoxic T cells. Class I HLA is expressed by the majority of nucleated cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| MHC I Free Chain | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| A4 | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| MHC Free | |
| Primary | |
| 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.