missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II RT1B Antibody (OX-6) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-65541JF585
This item is not returnable.
View return policy
Description
MHC Class II RT1B Monoclonal antibody specifically detects MHC Class II RT1B in Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunohistochemistry Free-Floating,CyTOF,Immunocytochemistry/ Immunofluorescence
Specifications
| MHC Class II RT1B | |
| Monoclonal | |
| Janelia Fluor 585 | |
| DMA, DMB, DP beta 1, DPB 1, DPB1, DRB, HLA class II histocompatibility antigen DM beta chain, HLA DMB, HLA DPB1, HLADM, HLADP1B, Major histocompatibility complex class II, Major histocompatibility complex class II DP beta 1, MHC class II antigen DMB, MHC DPB1, RING6, RING7 | |
| Rat thymocyte membrane glycoproteins | |
| 0.25 mL | |
| Adaptive Immunity, Immunology | |
| 24738 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), CyTOF, Immunocytochemistry/Immunofluorescence | |
| OX-6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction