missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II Rat, FITC, Clone: M5/114.15.2, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB70-F-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942388

  • £191.00 / 500µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MHC Class II
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
H2-Ab1 and H2-Eb1
Protein A/G Purified
RUO
14969
IgG2b κ
Flow Cytometry
M5/114.15.2
FITC
NP_034512
Rat
500 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers