missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Polymorphic Antibody (CI.13.I), Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £381.00
Specifications
| Antigen | MHC Class II Polymorphic |
|---|---|
| Clone | CI.13.I |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18082826
|
Novus Biologicals
NB100-64962-0.025ML |
0.025 mL |
£150.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18088454
|
Novus Biologicals
NB100-64962 |
0.125 mL |
£381.00
125µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MHC Class II Polymorphic Monoclonal specifically detects MHC Class II Polymorphic in Guinea Pig samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| MHC Class II Polymorphic | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Guinea Pig | |
| 3123 | |
| Guinea pig peritoneal T-cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CI.13.I | |
| Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Immunology, Major Histocompatibility Complexes | |
| Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
| HLA-DRB1 | |
| IgG2b | |
| Tissue culture supernatant | |
| Reacts with the MHC Class II epitope present on strain 13, but not strain 2 guinea pig cells. The antigen is also found in outbred Hartley guinea pigs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title