Learn More
Invitrogen™ MHC Class II (I-A/I-E) Recombinant Rat Monoclonal Antibody (P7/7)

Rat Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551851
Description
This antibody recognises the rat RT1Bu/Du MHC class II antigen, and cross-reacts with haplotypes c, d, l and n. This antibody stains MHC class II molecules in a number of rat strains, including PVG, F334, LEW, and BN. However, this antibody is not reactive with RT1a haplotype rats, including Wistar rats. This antibody also cross-reacts with murine I-A and I-E molecules of all haplotypes tested, including H-2b, d, f and k, so can be used in all mouse strains.
MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| MHC Class II (I-A/I-E) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P01910, P01921, P04227, P04228, P04230, P06342, P06343, P06344, P06345, P06346, P14434, P14435, P14436, P14437, P14438, P14483, P23150 | |
| H2-Aa, H2-Ab1, H2-Eb1 | |
| This antibody was raised by immunising DA rats with PVG.R8 rat spleen cells. Splenocytes from immunised rats were then isolated, and fused to the myeloma line NSO/u to generate stable hybridomas. | |
| 200 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| P7/7 | |
| Unconjugated | |
| H2-Ab1 | |
| Abeta; AI323765; AI845868; cell surface glycoprotein; E-alpha-f; H-2 class II histocompatibility antigen, A beta chain; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2 class II histocompatibility antigen, I-E beta chain; H-2Ab; H2-Ab; H2-Ab1; H-2Ea; H2-Ea; H2-Ea-ps; H2Eb; H-2Eb; H2-Eb1; H2-iabeta; H2-IE-alpha; histocompatibility 2, class II antigen A, beta 1; histocompatibility 2, class II antigen E alpha; histocompatibility 2, class II antigen E alpha, pseudogene; histocompatibility 2, class II antigen E beta; Ia2; Ia-2; Ia3; Ia-3; Ia4; Ia-4; IAb; I-Ab; I-Abeta; I-Ad; I-Aq; I-E alpha MHC class II; I-E beta MHC class II; I-Ealpha; I-Ed; I-Ek; major histocompatibility complex class II beta chain; major histocompatibility complex H-2E beta chain; MHC class 2; MHC class II antigen A beta; MHC class II antigen E alpha; MHC class II H2-IA-beta-psi; MHC class II IE-b; MHC class II protein; MHC H2-IE-beta cell surface glycoprotein; response to metastatic cancers 1; Rmcs1 | |
| Rat | |
| Protein A | |
| RUO | |
| 14960, 14961, 14969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.